Learn More
Andwin ScientificSupplier Diversity Partner AUTOCLAVE BAGS 48X58 200/CS  DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 14220040
Made with high-quality polypropylene, the Autoclavable Bags are made to securely hold nonhazardous waste. The bags can resist tears and autoclaving temperatures up to 135C (275F). This product complies with the strength requirements of the 165g dar
Catalog No. NC9103244