Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Autoclave Bags Printed with Biohazard Logo

Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.

Manufacturer:  Corning™ SA2110

 View more versions of this product

Catalog No. 07-202-046


Add to cart

Description

Description

  • Bags are made of 40µm thick transparent polypropylene
  • Wide range of volume available from 3L to 33L
Specifications

Specifications

780mm
21L
400mm
40μm
PP (Polypropylene)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit