Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves
SCI_ED

Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).

$52.25 - $59.65

Specifications

Certifications/Compliance CE
Color Orange
For Use With (Application) Handling Hot Objects
Material Terrycloth
Series H13201
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Cuff Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Cuff Price Quantity & Availability  
S47334
SCI_ED
View Documents
Bel-Art™
H132010000/EMD
5 in.
Pair for $59.65
Add to cart
 
S473341
SCI_ED
View Documents
Bel-Art™
H132010001/EMD
11 in.
Pair for $52.25
Add to cart
 
Description

Description

  • Ideal for removing hot objects from an autoclave or oven
  • Heat-resistant up to 450°F (232°C)
  • Replaces hazardous asbestos gloves
  • Extended gauntlet cuff available for additional forearm protection
  • Gloves are not waterproof and should not be used to handle dry ice
  • Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer
Specifications

Specifications

CE
Handling Hot Objects
H13201
Autoclave Gloves
Orange
Terrycloth
450°F
One Size
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit