Learn More
Bachem Autoinducing Peptide I, 0.5mg, 200010-29-3, Trifluoroacetate salt DFS Item
Manufacturer:  Bachem 4059504.0500
Autoinducing peptide (AIP) activating the agr (accessory gene regulator) locus to control the expression of extracellular proteins in S. aureus.
Catalog No. 50-194-7514