Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arlington Scientific Automated RPR Test Kit for Syphillis
Qualitative, nontreponemal, flocculation test for the detection of reagin antibodies in human serum and plasma as a screening test for serological evidence of syphilis for use on the Arlington Scientific Evolution analyzer. For use in diagnostic and blood donor screening.
Manufacturer: Arlington Scientific 9004800A
This item is not returnable.
View return policy
Specifications
4800 Tests | |
Serum and Plasma | |
24 Months from DOM | |
2°C to 8°C | |
86592 | |
Reagin | |
4800 | |
24 Months from DOM |
Test Kit | |
>99% | |
>99% | |
Self Declared | |
RPR test for Syphilis | |
Carbon antigen and test plates | |
110 µL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title