Learn More
Arlington Scientific Automated RPR Test Kit for Syphillis
Qualitative, nontreponemal, flocculation test for the detection of reagin antibodies in human serum and plasma as a screening test for serological evidence of syphilis for use on the Arlington Scientific Evolution analyzer. For use in diagnostic and blood donor screening.
Specifications
CE Marker Self declared
Clia Complexity Moderate
CPT Code 86592
Description RPR test for Syphilis
Detectable Analytes Reagin
View More Specs
Products 2
Catalog Number Mfr. No. Kit Size Price Quantity & Availability  
Catalog Number Mfr. No. Kit Size Price Quantity & Availability  
22-415-222
View Documents
Arlington Scientific
900480A
480 Tests Pack of 480 for $813.49
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-415-223
View Documents
Arlington Scientific
9004800A
4800 Tests Pack of 4800 for $7,802.97
N/A
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
Self declared
86592
Reagin
Test kit
110 µL
24 Months from DOM
2°C to 8°C
Moderate
RPR test for Syphilis
Carbon antigen and test plates
Carbon antigen
>99%
>99%
7 Minutes