Description
The membrane device automatically removes the aseptic packaging of the filter membrane, even available model with a touch-free mode induced by an optical sensor. No manual operation, free the user's hands, the membrane is distributed in the process, do not contact with other items, to avoid the risk of contamination.
- Stainless steel body spray processing, small size, beautiful shape
- Available with Power Supply or lithium-ion batteries
- AC drive can be connected to the charger
- Automatic distribution filter membrane
- Available model with optical sensor to take the film, without manual operation touch the button to take the film
- Fast and reliable transfer of filter membrane, filter membrane transfer by reel drive technology
- The filter membrane is easy to load
- Automatic collection of protective packaging
- Original color calibration technology, accurate detection
Specifications
Specifications
| Type | Membrane Dispenser |
| For Use With (Equipment) | Membrane Filters |