Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems

Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Manufacturer: Thermo Scientific™ 985261

 View more versions of this product

Catalog No. 14-754-487

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Color Clear
    For Use With (Equipment) BioTek Pipetting Systems
    Product Line BioTek
    Filtered No
    Sterility Sterile
    Volume (Metric) 100μL,100µL
    Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case,96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
    Sterile Yes
    Features Low retention
    Item Description 100uL; Sterile; Low retention
    Tip Format Low retention

    Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.  Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.

    • Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
    • Lot-tested during production on actual workstation for which they are created
    • Low percentage coefficient of variation (%CV)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit