Learn More
Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
$95.86
Specifications
For Use With (Equipment) BioTek™ Pipetting System
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Color Sterility Filtered Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Color Sterility Filtered Price Quantity & Availability  
14-754-483
View Documents
Thermo Scientific™
984021
50 μL Clear Sterile Filtered
Pack of 960 for $95.86
Case of 5 PK for $479.32
Add to cart
14-754-490
View Documents
Thermo Scientific™
986261
200 μL Clear Sterile Non-filtered This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.  Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
Specifications
BioTek™ Pipetting System
SDS
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.