Learn More
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations Available on GSA/VA Contract for Federal Government customers only.
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. 
Manufacturer:  Thermo Scientific™ 921262
 View more versions of this product
Catalog No. 21-402-305
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency.  Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
  • Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
Description & Specifications
Specifications
100μL
Nonsterile
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Caliper Zymark
No
Clear
Caliper Zymark Liquid Handling Workstations
Promotions
Promotions
  • Thermo Scientific
    Buy any three cases of Thermo Scientific™ Automation Tips and get a case of Automation Reservoirs at no additional cost.
    07-14-2017