Thermo Scientific™ Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Head Available on GSA/VA Contract for Federal Government customers only.

Increase pipetting performance with Thermo Scientific™ Automation Tips for  Perkin Elmer™/Packard™ Robotic Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Manufacturer: Thermo Scientific™ 925262

 View more versions of this product

Catalog No. 21-402-634

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Color Clear
    For Use With (Equipment) Packard Robotic Liquid Handling Systems
    Volume (Metric) 20μL,20µL
    Product Line PerkinElmer/Packard
    Sterility Nonsterile
    Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case,96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
    Filtered No
    Sterile No
    Features Low retention
    Item Description 20uL; Non-Sterile; Low retention
    Tip Format Low retention

    Increase pipetting performance with Thermo Scientific™ Automation Tips for  Perkin Elmer™/Packard™ Robotic Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.  Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: PerkinElmer/Packard PlateTrak, PerkinElmer/Packard MiniTrak, PerkinElmer Evolution P-3 and PerkinElmer JANUS;  Molecular Device FLIPR with Type "A" CCS 384-well head and Type "B" Liberty 384-well head.

    • Available in non-sterile, sterile, with Thermo Scientific™ ART™ filter technology and in low retention
    • Increased precision for the toughest multi-dispensing assays
    • Low percentage coefficient of variation (%CV)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit