Learn More
Mopec Autopsy Table Sink Service with Cabinet Available on GSA/VA Contract for Federal Government customers only.
Mopec™ Autopsy Table Sink Service with Cabinet accommodates all Mopec™ autopsy carts and includes all amenities to perform an efficient autopsy.
Manufacturer:  Mopec CE850
Catalog No. 22-444-035
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Tabletop is 14 gauge stainless steel with #4 satin finish
  • Table Wing is fabricated of 18 gauge stainless steel
  • 2 GFCI Receptacles positioned on each side of sink
  • Hydro aspirator located on the end of the table above the sink basin
  • Provided with a reversing valve to clear the aspirator of any foreign matter
  • Completely plumbed with control valve, vacuum breaker and tubing
  • Hot and cold single lever mixing valve with swing spout faucet complete with vacuum breaker
  • Deck mounted chrome plated brass sink spray with high pressure hose
  • Table wing includes a medium size instrument drawer and storage compartment
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
1 Year Warranty
Specifications
Specifications
2 GFCI Receptacles positioned on each side of sink
Autopsy Tables
Faucet and sink spray