Learn More
Azzota Corp AUY-922 DFS Item
Manufacturer:  Azzota Corp C1239
AUY-922 (Luminespib) is a derivative of 4,5-diarylisoxazole and a third-generation heat shock protein 90 (Hsp90) inhibitor with potential antineoplastic activity. Please request if need more quantities
Catalog No. 50-161-3155