Learn More
Scott Safety™ AV-3000™ Facepieces
Improve on the popular AV-2000™ facepiece
Manufacturer:  Scott Safety™ 80577302
 View more versions of this product
Catalog No. 19-148-614
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : Requires adapter SCT80405701.
Description
Description
Full-facepiece assembly fits persons of varying facial shapes and sizes with minimal interference to downward and peripheral vision. With appropriate adapter (SCT80405701), the AV-3000 can also be used as an air-purifying respirator with 42CFR84 cartridges.
  • Polycarbonate lens meets the faceshield impact and penetration requirements of ANSI Z87.1 paragraphs 5.2.8.1 and 5.2.8.2, and the lens abrasion requirements of NFPA1981
  • Comfort Seal™ faceseal with a large sealing area for a better hood-to-facepiece interface
  • Easy-to-don, six-point, quad-suspension net hood
  • Larger multidirectional voicemitters provide enhanced clarity in voice communication
  • Simplistic clam shell design for decreased part changeout
Certifications
NIOSH approved, NFPA 2007 approved.
Specifications
Specifications
NIOSH, NFPA 2007
Facepiece
Used as an air purifying respirator
Larger multidirectional voicemitters
Dupont™ Kevlar™ aramid fiber, Polyisoprene Seal
Medium
42cfr84 cartridge