Learn More
Scott Safety™ AV-3000™ Facepieces
Improve on the popular AV-2000™ facepiece
Supplier: Scott Safety™ 80577303
Requires : Requires adapter SCT80405701.
Description
Full-facepiece assembly fits persons of varying facial shapes and sizes with minimal interference to downward and peripheral vision. With appropriate adapter (SCT80405701), the AV-3000 can also be used as an air-purifying respirator with 42CFR84 cartridges.
- Polycarbonate lens meets the faceshield impact and penetration requirements of ANSI Z87.1 paragraphs 5.2.8.1 and 5.2.8.2, and the lens abrasion requirements of NFPA1981
- Comfort Seal™ faceseal with a large sealing area for a better hood-to-facepiece interface
- Easy-to-don, six-point, quad-suspension net hood
- Larger multidirectional voicemitters provide enhanced clarity in voice communication
- Simplistic clam shell design for decreased part changeout
Certifications
NIOSH approved, NFPA 2007 approved.
Specifications
NIOSH, NFPA 2007 | |
Facepiece | |
Used as an air purifying respirator | |
Larger multidirectional voicemitters |
Dupont™ Kevlar™ aramid fiber, Polyisoprene Seal | |
Large | |
42cfr84 cartridge |