Learn More
Tocris™ AVE 1625
Potent and selective CB1 receptor antagonist
Manufacturer:  Tocris™ 6763/10
 View more versions of this product
Catalog No. 67-631-0
Add to cart
This item is not returnable. View return policy
Description
Description
AVE 1625 is a potent and selective CB1 receptor antagonist (IC50 values are 25 and 10 nM for human and rat CB1R, respectively). Exhibits >400-fold selectivity for CB1 over CB2 receptors. Attenuates Olanzapine (Cat. No. 4349) induced weight gain and food intake in rats with no effect on energy expenditure or motility. Also improves cognitive function in rodent schizophrenia models and ameliorates extrapyramidal side-effects of antipsychotics.
Specifications
Specifications
497.38
Potent and selective CB1 receptor antagonist
Drinabant
C23H20Cl2F2N2O2S
98%
10 mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C