Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avi Tag Mouse, Clone: 1D11D10, GenScript™

Catalog No. 502120580
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0580 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0580 Supplier Genscript Corporation Supplier No. A0173840
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. Avi tag is a biotin-acceptor peptide, GLNDIFEAQKIEWHE. The 15-residue peptide served as a substrate mimic for biotin ligase (BirA), which usually recognizes the much larger protein domain. Avi Tag Antibody is a useful tool in analysis of Avi fusion proteins.

Avi Tag Monoclonal antibody specifically detects Avi Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen Avi Tag
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1D11D10
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias Avi; BirA
Host Species Mouse
Immunogen Avi peptide conjugated to KLH
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2a κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.