Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian avulavirus 1 Fusion Glycoprotein F0, Rabbit anti-Virus, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP280440
Description
Avian avulavirus 1 Fusion Glycoprotein F0 Polyclonal antibody specifically detects Avian avulavirus 1 Fusion Glycoprotein F0 in Virus Samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin).Specifications
Avian avulavirus 1 Fusion Glycoprotein F0 | |
1 mg/mL | |
Western Blot 1:500-1:1000, ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin | |
Affinity Purified | |
Synthetic peptide corresponding to Avian avulavirus 1 Fusion glycoprotein F0 aa 540-554. | |
Peptide affinity purified | |
RUO | |
Primary | |
Virus |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
1X TBS(pH7.4), 0.5%BSA, 50%Glycerol | |
Rabbit | |
IgG | |
100 uL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Polyclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title