Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Avian Clusterin/APOJ ELISA Kit (Colorimetric)

Useful in the study of cancer markers.

$602.00

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 7.813 to 500ng/mL
Species Avian
Sensitivity 4.963ng/mL
Sample Type Serum, plasma and tissue homogenates
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599841KT View Documents Novus Biologicals™
NBP2599841KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Avian Clusterin/APOJ ELISA Kit (Colorimetric) measures avian Clusterin/APOJ in serum, plasma and tissue homogenates.
Specifications

Specifications

ELISA Kit (Colorimetric)
Avian
Serum, plasma and tissue homogenates
Sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin, TRPM-2TRPM2
Clusterin/APOJ
7.813 to 500ng/mL
4.963ng/mL
RUO
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit