Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Avian Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin ELISA Kit (Colorimetric)
Recognizes both recombinant and natural H9N2 Avian Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin.
Manufacturer: Novus Biologicals NBP280342
Specifications
78.13 to 5000 pg/mL | |
6.9 pg/mL | |
1 Kit | |
Recombinant protein |
ELISA Kit (Colorimetric) | |
Storage is content dependent | |
Avian Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title