Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AvianInfluenzaAHemagglutinin Mouse anti-Influenza virus, Clone: 8J159, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266608

Catalog No. NBP266608


Add to cart

Description

Description

AvianInfluenzaAHemagglutinin Monoclonal antibody specifically detects AvianInfluenzaAHemagglutinin in Influenza virus samples. It is validated for ELISA, Haemagglutination, Dot Blot.
Specifications

Specifications

AvianInfluenzaAHemagglutinin
8J159
ELISA 1:100-1:2000, Haemagglutination, Dot Blot 1:100-1:2000
Purified
Mouse
IgG2a
100μg
Primary
Influenza virus
Dot Blot, ELISA, Hemagglutination Assay
Unconjugated
PBS (pH 7.4) with 0.1% Sodium Azide
ha; HA1; HA2; Hemagglutinin; Hemagglutinin ha1 chain; Hemagglutinin ha2 chain; Hemagglutinin precursor
Hybidoma from Sp2/0 myeloma cells with spleen cells from Balb/c mice immunized with purified Avian Influenza Virus Type A (H5N1).
Protein G purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit