Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Avidin from Egg White
Avidin from Egg White, 10-15 u/mg solid, White Crystalline Powder
$122.00 - $1032.00
Specifications
Color | White |
---|---|
Source | Egg White |
CAS | 1405-69-2 |
Content And Storage | -20°C |
Form | Crystalline |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP215004705
|
MP Biomedicals™
0215004705 |
5 mg |
Each for $122.00
|
|
|||||
MP215004710
|
MP Biomedicals™
0215004710 |
10 mg |
Each for $216.00
|
|
|||||
MP215004725
|
MP Biomedicals™
0215004725 |
25 mg |
Each for $372.00
|
|
|||||
MP215004780
|
MP Biomedicals™
0215004780 |
100 mg |
Each for $1,032.00
|
|
|||||
Description
- Avidin is useful in a biotin-avidin system for labeling biomolecules for receptor studies, immunoassays and immunohistological methods.
- Avidin egg white was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding.
- Egg white was used at 80 nmol.
- Egg white avidin has been used as a cross-linker for biotin-conjugated antibodies.
- The product has also been used as a blocking agent in ELISA applications.
- Avidin is a basic glycoprotein isolated from raw egg white.
- It exhibits high binding affinity for biotin and is capable of producing biotin deficiency in rats and chicks.
- It also occurs in the white portion of eggs and the oviducts of birds and amphibia.
- It is destroyed by cooking or irradiation.
- One unit will bind 1 ug of D-biotin at pH 8.9.
Specifications
White | |
1405-69-2 | |
Crystalline |
Egg White | |
-20°C | |
Labeling biomolecules for receptor studies, immunoassays and immunohistological methods |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title