Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AVIL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568521
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 93%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Specifications
AVIL | |
Polyclonal | |
Unconjugated | |
AVIL | |
actin-binding protein DOC6; Advil; Advillin; Advillin-like protein; Avil; DOC6; p92; peripheral nervous system villin-like protein; pervin; Unknown (protein for MGC:136857); zgc:136857 | |
Rabbit | |
Affinity chromatography | |
RUO | |
79253 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9WU06 | |
AVIL | |
synthetic peptide directed towards the middle region of human AVIL. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction