Learn More
Medchemexpress LLC Avobenzone | 70356-09-1 | 98.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0316500MG
Encompass_Preferred
Avobenzone, a dibenzoylmethane compound, is a widely used filter in sunscreens for skin photoprotection in the UVA band. It is also identified as an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.
  • Acts as a filter in sunscreens for UVA skin photoprotection.
  • Functions as an endocrine disruptor by binding to estrogen receptor β.
  • Promotes adipogenesis in human bone marrow mesenchymal stem cells.
  • Up-regulates mRNA levels of PPARγ during adipogenesis in human bone marrow mesenchymal stem cells.
  • Inhibits proliferative activities of human trophoblast cells.
  • Induces apoptosis in HTR8/SVneo cells.
  • Exhibits weak ERα agonism and weak AR antagonism.
Catalog No. 50-163-9324