Learn More
AVPI1 Mouse anti-Human, Clone: OTI6C5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802991
Catalog No. 50-166-6281
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be involved in MAP kinase activation, epithelial sodium channel (ENaC) down-regulation and cell cycling.
Specifications
Specifications
AVPI1
Monoclonal
Unconjugated
AVPI1
Lyophilized
AVPI1
Full length human recombit protein of human AVPI1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6C5
PBS with 8% trehalose and no preservative; pH 7.3
Q5T686
PP5395, VIP32, VIT32
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
60370