Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AWAT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550948

Catalog No. PA550948


Add to cart

Description

Description

AWAT1 Polyclonal Antibody detects endogenous levels of AWAT1 protein.

The protein encoded by this gene belongs to the diacylglycerol acyltransferase family. It esterifies long chain (wax) alcohols with acyl-CoA-derived fatty acids to produce wax esters. Wax esters are enriched in sebum, suggesting that this enzyme plays a central role in lipid metabolism in skin. Consistent with this observation, this protein is predomitly expressed in the sebaceous gland of the skin. [provided by RefSeq, Sep 2009].
Specifications

Specifications

AWAT1
Polyclonal
Unconjugated
AWAT1
Liquid
AWAT1
Synthesized peptide derived from the C-terminal region of human DGAT2L3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q58HT5
DGA2, DGAT2L3
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
158833
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit