Learn More
AWAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 251191AP
Catalog No. 50-173-4733
Add to cart
Description
Description
This gene encodes an enzyme belonging to the diacylglycerol acyltransferase family. This enzyme produces wax esters by the esterification of long chain (or wax) alcohols with acyl-CoA-derived fatty acids. It functions in lipid metabolism in the skin, mostly in undifferentiated peripheral sebocytes. This enzyme may also have acyl-CoA:retinol acyltransferase activities, where it catalyzes the synthesis of diacylglycerols and retinyl esters.
Specifications
Specifications
AWAT2
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6E1M8, Q6E213
AWAT2, DC4, DGAT2L4, hDC4, hWS, MFAT, Wax synthase, WS
Rabbit
IgG
150 μL
-20°C
Primary
158835, 245532, 302425
Western Blot
Unconjugated
AWAT2
Liquid
Awat2
AWAT2 Fusion Protein Ag18936
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat