Learn More
Abnova™ AXL 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000558T01
Catalog No. 89-936-217
Add to cart
Description
Description
Specifications
Specifications
293T
BC032229.1
pCMV-AXL full-length
100 μL
Human
AXL 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 98.45kDa
Transient overexpression cell lysate was tested with anti-AXL antibody (Mfr. No. H00000558-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot