Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axl Rat anti-Mouse, Alexa Fluor 647, Clone: 100000000, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11094151AF647

Catalog No. NBT94151647


Add to cart

Description

Description

Axl Monoclonal antibody specifically detects Axl in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Axl
100000000
Western Blot
Purified
AXL
Recombinant Mouse Axl extracellular domain
Protein G purified
Cell Cycle and Replication, Phospho Specific
Primary
558
Mouse
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This recognizes mouse Axl in western blot. It has no cross reactivity to rmMer or rmDtk.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit