Learn More
Alfa Aesar™ Axltide
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66170EXD
 View more versions of this product
Catalog No. AAJ66170EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1514.78
0.5mg
Lys-Lys-Ser-Arg-Gly-Asp-Tyr-Met-Thr-Met-Gln-Ile-Gly
-30° to -10°C
Axltide
HPLC
Solid
RUO
Axl/Insulin Receptor substrate peptide