Learn More
Axygen™ 0.5mL Conical Screw Cap Tubes: Nonsterile
Features chemical resistant O-ring
Manufacturer:  Axygen Scientific SCT050A
 View more versions of this product
Catalog No. 14-222-516
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
RNase, DNase, pyrogen-free
Polypropylene
0.5mL
20000xG
Non-looped
O-ring
Conical
Assorted
Screw
Polypropylene
No
No
Bag