Axygen™ 0.5mL Conical Screw Cap Tubes: Nonsterile Available on GSA/VA Contract for Federal Government customers only.
Features chemical resistant O-ring
Manufacturer: Axygen Scientific SCT050C
 View all options for this product
Catalog No. 14-222-522
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Certifications/Compliance RNase, DNase, pyrogen-free
    Closure Color Clear
    Closure Material Polypropylene
    Closure Type Screw
    Capacity (Metric) 0.5mL
    Material Polypropylene
    Max. Force 20000xG
    Sterile No
    Format Non-looped
    Graduated No
    Includes O-ring
    Packaging 500 Tubes and caps/Pk., 8 Pk./Cs.
    Shape Conical
    Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
    • Nonsterile cap tubes have unattached, non-looped caps with O-rings
    • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
    • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
    • Available in range of colors for easy sample identification
  • Description & Specifications
Related Products
Product Content Feedback