Axygen™ 0.5mL Conical Screw Cap Tubes: Nonsterile
Features chemical resistant O-ring
Manufacturer: Axygen Scientific SCT050V
 View more versions of this product
Catalog No. 14-222-556
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Capacity (Metric) 0.5mL
    Material Polypropylene
    Max. Force 20000xG
    Sterile No
    Format Non-looped
    Graduated No
    Includes O-ring
    Packaging 500 Tubes and caps/Pk., 8 Pk./Cs.
    Certifications/Compliance RNase, DNase, pyrogen-free
    Shape Conical
    Closure Color Violet
    Closure Material Polypropylene
    Closure Type Screw
  • Description & Specifications
Related Products
Product Content Feedback