Axygen™ 0.5mL Self Standing Screw Cap Tubes Available on GSA/VA Contract for Federal Government customers only.
Features O-ring manufactured for resistant to many commonly-used laboratory chemicals
Manufacturer: Axygen Scientific SCT050SSB
 View more versions of this product
Catalog No. 14-222-535
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance RNase, DNase, pyrogen-free
    Closure Color Blue
    Closure Material Polypropylene
    Closure Type Screw
    Capacity (Metric) 0.5mL
    Material Polypropylene
    Max. Force 20000xG
    Sterile No
    Format Non-looped
    Graduated No
    Includes O-ring
    Shape Conical
    For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
    • Available as presterilized and nonsterile tubes
    • Nonsterile cap tubes have unattached, non-looped caps with O-rings
    • Presterilized cap tubes have non-looped caps attached with O-rings
    • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
    • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
    • Wide range of colored tubes for easy sample identification
  • Description & Specifications
Related Products
Product Content Feedback