Axygen™ 1.5mL Self-Standing Screw Cap Tubes Available on GSA/VA Contract for Federal Government customers only.
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
Manufacturer: Axygen Scientific SCT150SSRS
 View more versions of this product
Catalog No. 14-222-594
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 1.5mL
    Material Polypropylene
    Max. Force 20000xG
    Sterile Yes
    Format Non-looped
    Includes O-ring
    Certifications/Compliance RNase, DNase, pyrogen-free
    Shape Conical
    Closure Color Red
    Closure Material Polypropylene
    Closure Type Screw
  • Description & Specifications
Related Products
Product Content Feedback