Axygen™ 1.5mL Self-Standing Screw Cap Tubes
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
Manufacturer: Axygen Scientific SCT150SSO
 View more versions of this product
Catalog No. 14-222-590
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Capacity (Metric) 1.5mL
    Material Polypropylene
    Max. Force 20000xG
    Sterile No
    Format Non-looped
    Graduated No
    Includes O-ring
    Certifications/Compliance RNase, DNase, pyrogen-free
    Shape Conical
    Closure Color Orange
    Closure Material Polypropylene
    Closure Type Screw
  • Description & Specifications
Related Products
Product Content Feedback