Axygen™ 1.5mL Self-Standing Screw Cap Tubes Available on GSA/VA Contract for Federal Government customers only.
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
Manufacturer: Axygen Scientific SCT150SSR
 View more versions of this product
Catalog No. 14-222-593
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Capacity (Metric) 1.5mL
    Material Polypropylene
    Max. Force 20000xG
    Format Non-looped
    Graduated No
    Includes O-ring
    Certifications/Compliance RNase, DNase, pyrogen-free
    Closure Color Red
    Closure Material Polypropylene
    Closure Type Screw
    Sterile No
    Shape Conical
  • Description & Specifications
Related Products
Product Content Feedback