Axygen™ 12-Strip PCR Tubes and Caps Available on GSA/VA Contract for Federal Government customers only.

Manufactured from 99.9% pure virgin polypropylene

Manufacturer: Axygen Scientific PCR0212FCPC

 View more versions of this product

Catalog No. 14-222-259

  Check Availability
Add to Cart

    Specifications

    Type Thin Wall 12-Strip PCR Tube And Flat Cap
    Volume (Metric) 0.2mL
    Quantity 80 Pack
    Sterile No
    Closure Material Polypropylene
    Max. Force 4000xG
    Cap Type Flat
    Format Strip
    For Use With (Equipment) Thermal cyclers for thermal transfer applications
    Capacity (Metric) 0.2mL
    Certifications/Compliance RNase, DNase-free
    Color Clear
    Material Polypropylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit