Axygen™ 1250µL Universal Pipette Tips

Matrix compatible tips in clear color

Manufacturer: Axygen Scientific MTX1250CL

 View more versions of this product

Catalog No. 14-223-311

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Matrix-style, Maxymum Recovery, Bevelled
    Capacity (Metric) 1250μL
    Color Clear
    Material Polypropylene
    Sterile No
    For Use With (Equipment) Matrix Impact™ pipettors
    Packaging Bulk
    Certifications/Compliance RNase, DNase-free
    Autoclavable Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit