Learn More
Axygen™ 20µL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format Available on GSA/VA Contract for Federal Government customers only.
For use with Agilent/Velocity11 automation systems
Manufacturer:  Axygen Scientific VT20RS
 View more versions of this product
Catalog No. 14-222-878
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Robotic Tip
Sterile
Racked
Polypropylene
96 Tips/Rack
Non-pyrogenic, used for stringent specifications under strict process controls
20μL
Clear
Round
Rack
RNase, DNase-free, ANSI
Agilent™/Velocity11™ automated liquid handling system and VPrep™ and Bravo™ 96LT and 8LT head