Axygen™ 20µL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format

For use with Agilent/Velocity11 automation systems

Manufacturer: Axygen Scientific VT20RLAB

 View more versions of this product

Catalog No. 14-222-876

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Robotic Tip
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Agilent™/Velocity11™ automated liquid handling system and VPrep™ and Bravo™ 96LT and 8LT head
    Sterility Nonsterile
    Tip Style Racked
    Shape Round
    Packaging Rack
    Certifications/Compliance RNase, DNase-free, ANSI
    For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
    Volume (Metric) 20μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit