Learn More
Axygen™ 200µL Universal Pipette Tips200µL, Beveled, Reference Available on GSA/VA Contract for Federal Government customers only.
Designed to fit a wide variety of single and multi-channel pipettes
Manufacturer:  Axygen Scientific TR222YRS
 View more versions of this product
Catalog No. 14-222-822
  Check Availability
Add to cart
Specifications
Description
  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
  • Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
Description & Specifications
Specifications
Universal Pipetter Tips
Sterile
10, 50, 100μL
Beveled
Rack
Wide variety of single and multichannel pipetters
1 to 200μL
Yellow
Yes
Polypropylene
DNase, RNase, pyrogen free