Axygen™ 200µL Universal Pipette Tips200µL, Beveled, Reference Available on GSA/VA Contract for Federal Government customers only.

Designed to fit a wide variety of single and multi-channel pipettes

Manufacturer: Axygen Scientific TR222CLRS

 View more versions of this product

Catalog No. 14-222-814

  Check Availability
Add to Cart

    Specifications

    Product Type Universal Pipetter Tips
    Material Polypropylene
    For Use With (Equipment) Wide variety of single and multichannel pipetters
    Sterility Sterile
    Graduations 10, 50, 100μL
    Autoclavable Yes
    Tip Style Beveled, Maxymum Recovery
    Packaging Rack
    Certifications/Compliance DNA, DNase, RNase and PCR inhibitor-free
    Volume (Metric) 1 to 200μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit