Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ 24- and 32-Well PCR Microplates, Clear, Nonsterile
Designed to fit a wide variety of thermal cyclers
Supplier: Axygen™ PCR32C
Description
- Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
Specifications
RNase, DNase-free | |
Clear | |
Polypropylene | |
Non-sterile | |
PCR | |
10/Pk. |
Without Lid | |
PCR Microplate, without Lid | |
32 | |
Round | |
50/Cs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction