Learn More
Axygen™ 300µL Universal Pipetter Tips, Maxymum Recovery Available on GSA/VA Contract for Federal Government customers only.
Maxymum Recovery tips fit popular brands of pipettes
Manufacturer:  Axygen Scientific T350CL
 View more versions of this product
Catalog No. 14-222-747
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Universal Tip
Nonsterile
Maxymum Recovery™
Bulk
RNase, DNase, and endotoxin free
1 to 300μL
Clear
Polypropylene
10000 Case
Single and multichannel pipettors