Axygen™ 384-Well PCR Half-Skirt Microplates Available on GSA/VA Contract for Federal Government customers only.

Optimized for MJ Research Cyclers

Manufacturer: Axygen Scientific PCR384M2CBC

 View more versions of this product

Catalog No. 14-223-072

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    For Use With (Application) For use within PCR thermal transfer and RT-PCR applications
    For Use With (Equipment) For ABI 3730, 3100 and MJ thermocyclers
    Volume (Metric) Working 50μL
    Color Clear
    Product Type 384-Well PCR Microplate
    Applications For use within PCR thermal transfer and RT-PCR applications

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit