Learn More
Axygen™ 384-well Skirted PCR Microplates
Single and double notch 384-well PCR plates
Manufacturer:  Axygen Scientific PCR384Y
 View more versions of this product
Catalog No. 14-222-318
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Sealing Options: Axymats, heat sealing, adhesive films and foils.
  • Available in 50µL and 55µL
  • Single and double notch corner
Skirted (50µL)
  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55µL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50µL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55µL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Colors available by special order.
Description & Specifications
Specifications
RNase, DNase-free
PCR
Polypropylene
No
40μL
Round
Yellow
Double notch
Full
40μL
Yellow