Axygen™ 384-well Skirted PCR Microplates

Single and double notch 384-well PCR plates

Manufacturer: Axygen Scientific PCR384M2SP

 View more versions of this product

Catalog No. 14-222-312

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) PCR, thermal transfer applications
    For Use With (Equipment) For ABI 3730, 3100 and MJ thermocyclers
    Includes Single notch
    Material Polypropylene,Polypropylene
    Skirt Style Full
    Sterile No,No
    Certifications/Compliance RNase, DNase-free
    Volume (Metric) Well 50μL
    Volume (Metric) Working 50μL
    Well Color Assorted
    Well Design Round
    Color Assorted
    Product Type 384-Well PCR Microplate
    Applications PCR, thermal transfer applications
    Well Shape Round

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit