Learn More
Axygen™ 50-well Axyracks
Ideal for cryogenic storage to -80°C and below
Manufacturer:  Axygen Scientific R50BK
 View more versions of this product
Catalog No. 14-223-349
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Black
Clear Polycarbonate base and cover
50.0
1.5 to 2mL Tubes
Polycarbonate
10/Pk.