Axygen™ 8-Strip PCR Tubes, 0.2mL Available on GSA/VA Contract for Federal Government customers only.

Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to prevent sample loss or without caps.

Manufacturer: Axygen™ PCR0208C

 View more versions of this product

Catalog No. 14-222-250

  Check Availability
Add to Cart

    Specifications

    Cap Type Sold separately
    Type Thin Wall 8-Strip PCR Tube
    Volume (Metric) 0.2mL
    Closure Material Polypropylene
    Max. Force 4000xG
    Format Strip
    For Use With (Equipment) Thermal cyclers for thermal transfer applications
    Shape Conical
    Capacity (Metric) 0.2mL
    Certifications/Compliance RNase, DNase, pyrogen-free
    Quantity 125 Pack
    Sterile No
    Color Clear
    Material Polypropylene

    • PCR tubes strips designed to fit a wide variety of thermal cyclers
    • Ultra-thin and consistent wall thickness allows precise thermal transfer
    • Domed or flat insertion caps ensure a tight fit that prevents sample evaporation
    • Certified RNase-/DNase-free and nonpyrogenic
    • Manufactured with 99.9% pure virgin polypropylene
    • Available in a variety of colors for easy sample identification

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit